Assay Details
Target Gene Details
Entrez Gene ID: | 268390 |
Gene Name: | AHA1, activator of heat shock protein ATPase 2 |
Gene Aliases: |
- |
Location: |
Chr.11:23487882-23498039 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ahsa2 | NM_001290654.1 | 1 | 176 | NP_001277583.1 |
| NM_001290655.1 | 1 | 176 | NP_001277584.1 | |
| NM_172391.4 | 1 | 176 | NP_765979.3 | |
| AK033696.1 | 1 | 84 | ||
| AK093945.1 | 1 | 83 | BAC04256.1 | |
| AK137116.1 | 1 | 87 | BAE23240.1 | |
| AK154196.1 | 1 | 87 | BAE32431.1 | |
| AK155265.1 | 1 | 87 | BAE33153.1 | |
| BC038397.1 | 1 | 67 | ||
| BC052829.1 | 1 | 94 | AAH52829.1 | |
| BC064012.1 | AAH64012.1 | |||
| BY013732.1 | 1 | 81 | ||
| BY023007.1 | 1 | 65 | ||
| CJ147821.1 | 1 | 177 |
More Information
Set Membership: |
|

Genomic Map