Assay Details
Target Gene Details
Entrez Gene ID: | 207165 |
Gene Name: | bromodomain PHD finger transcription factor |
Gene Aliases: |
9430093H17Rik, Falz |
Location: |
Chr.11:107033081-107131922 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bptf | NM_176850.2 | NP_789820.2 | ||
| AK143675.1 | BAE25490.1 | |||
| BC032289.1 | AAH32289.1 | |||
| BC060715.1 | AAH60715.1 | |||
| GU189417.1 |

Genomic Map