Assay Details
Target Gene Details
Entrez Gene ID: | 11370 |
Gene Name: | acyl-Coenzyme A dehydrogenase, very long chain |
Gene Aliases: |
vlcad |
Location: |
Chr.11:70010183-70015428 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acadvl | NM_017366.3 | 20 | 1978 | NP_059062.1 |
| AF017176.1 | 17 | 1626 | AAC31642.1 | |
| AI843782.1 | 1 | 228 | ||
| AK076037.1 | 20 | 1939 | ||
| AK146334.1 | 15 | 1451 | BAE27089.1 | |
| BC026559.1 | 20 | 1957 | AAH26559.1 | |
| U41497.1 | 11 | 860 | AAA85185.1 | |
| Z71189.1 | 20 | 1899 | CAA94919.2 |
More Information
Set Membership: |
|

Genomic Map