Assay Details
Target Gene Details
Entrez Gene ID: | 56697 |
Gene Name: | A kinase (PRKA) anchor protein 10 |
Gene Aliases: |
1500031L16Rik, B130049N18Rik, D-AKAP-2, D-AKAP2, PRKA10 |
Location: |
Chr.11:61871307-61930307 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akap10 | NM_019921.3 | NP_064305.2 | ||
| AK049399.1 | BAC33735.1 | |||
| AK083255.1 | ||||
| AK162966.1 | BAE37139.1 |
More Information
Set Membership: |
|

Genomic Map