Assay Details
Target Gene Details
Entrez Gene ID: | 216345 |
Gene Name: | zinc finger, C3H1-type containing |
Gene Aliases: |
BC033596, Ccdc131, Psrc2 |
Location: |
Chr.10:115384920-115432772 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfc3h1 | NM_001033261.2 | NP_001028433.2 | ||
| AK172969.1 | BAD32247.1 | |||
| BC033596.1 | AAH33596.1 | |||
| BC061510.1 | AAH61510.1 | |||
| BC139124.1 |

Genomic Map