Assay Details
Target Gene Details
Entrez Gene ID: | 14674 |
Gene Name: | guanine nucleotide binding protein, alpha 13 |
Gene Aliases: |
AU024132, AU043124, Galpha13 |
Location: |
Chr.11:109362783-109401369 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gna13 | NM_010303.3 | 2 | 579 | NP_034433.3 |
| AK011851.1 | 2 | 542 | BAB27880.1 | |
| AK149766.1 | 2 | 473 | BAE29071.1 | |
| BC057665.1 | 2 | 433 | AAH57665.1 | |
| M63660.1 | 2 | 527 | AAA37649.1 |
More Information
Set Membership: |
|

Genomic Map