Assay Details
Target Gene Details
Entrez Gene ID: | 14674 |
Gene Name: | guanine nucleotide binding protein, alpha 13 |
Gene Aliases: |
AU024132, AU043124, Galpha13 |
Location: |
Chr.11:109362783-109401369 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gna13 | NM_010303.3 | NP_034433.3 | ||
| AK149766.1 | BAE29071.1 | |||
| BC057665.1 | AAH57665.1 | |||
| M63660.1 | AAA37649.1 |
More Information
Set Membership: |
|

Genomic Map