Assay Details
Target Gene Details
Entrez Gene ID: | 14694 |
Gene Name: | guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1 |
Gene Aliases: |
AL033335, GB-like, Gnb2-rs1, Rack1, p205 |
Location: |
Chr.11:48800360-48806241 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnb2l1 | NM_008143.3 | NP_032169.1 | ||
| AK002493.1 | BAB22141.1 | |||
| AK012242.1 | BAB28114.1 | |||
| AK017772.1 | BAB30920.1 | |||
| AK051226.1 | BAC34564.1 | |||
| AK159797.1 | BAE35378.1 | |||
| AK160528.1 | BAE35846.1 | |||
| AK160739.1 | BAE35980.1 | |||
| AK161234.1 | BAE36257.1 | |||
| AK166417.1 | BAE38762.1 | |||
| AK166786.1 | BAE39017.1 | |||
| AK166800.1 | BAE39028.1 | |||
| AK166868.1 | 1 | 1451 | ||
| AK166889.1 | BAE39095.1 | |||
| AK166945.1 | BAE39133.1 | |||
| AK168086.1 | BAE40059.1 | |||
| AK168096.1 | BAE40068.1 | |||
| AK168196.1 | BAE40156.1 | |||
| AK168349.1 | BAE40286.1 | |||
| AK169612.1 | ||||
| BC046760.1 | AAH46760.1 | |||
| D29802.1 | BAA06185.1 | |||
| X75313.1 | CAA53062.1 |

Genomic Map