Assay Details
Target Gene Details
Entrez Gene ID: | 52626 |
Gene Name: | CDKN2A interacting protein N-terminal like |
Gene Aliases: |
A430101B06Rik, AA409260, D11Ertd497e, MNCb-1520 |
Location: |
Chr.11:51967631-51977336 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdkn2aipnl | NM_029976.3 | 2 | 379 | NP_084252.1 |
| AB041653.1 | 2 | 347 | BAB93546.1 | |
| AK020751.1 | 2 | 352 | BAB32201.1 | |
| AK077515.1 | 2 | 204 | ||
| AK150573.1 | 2 | 335 | BAE29668.1 | |
| AK171740.1 | 2 | 314 | BAE42643.1 | |
| BC116362.2 | 2 | 339 | ||
| BC116363.2 | 2 | 339 | ||
| BY088687.1 |
More Information
Set Membership: |
|

Genomic Map