Assay Details
Target Gene Details
Entrez Gene ID: | 67487 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 40 |
Gene Aliases: |
2410016C14Rik, ARG147, DDX40, PAD |
Location: |
Chr.11:86768849-86807660 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhx40 | NM_026191.2 | NP_080467.3 | ||
| AK004610.1 | BAC25091.1 | |||
| AK010512.1 | BAB26995.2 | |||
| AK029935.1 | ||||
| AK031000.1 | BAC27210.1 | |||
| AK135592.1 | ||||
| BC058276.1 | ||||
| BY329362.1 |
More Information
Set Membership: |
|

Genomic Map