Assay Details
Target Gene Details
Entrez Gene ID: | 18612 |
Gene Name: | ets variant 4 |
Gene Aliases: |
AW414408, Pea-3, Pea3 |
Location: |
Chr.11:101769742-101785310 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Etv4 | NM_008815.2 | NP_032841.2 | ||
| AK144538.1 | ||||
| AK144590.1 | BAE25952.1 | |||
| AK144683.1 | ||||
| AK158497.1 | BAE34538.1 | |||
| AK166136.1 | ||||
| BY731410.1 | ||||
| DQ832277.1 |
More Information
Set Membership: |
|

Genomic Map