Assay Details
Target Gene Details
Entrez Gene ID: | 216846 |
Gene Name: | centrobin, centrosomal BRCA2 interacting protein |
Gene Aliases: |
9830165K03Rik, Lip8 |
Location: |
Chr.11:69299491-69323893 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cntrob | NM_172560.3 | NP_766148.2 | ||
| AK034947.1 | ||||
| AK036710.1 | BAC29545.1 | |||
| AK147774.1 | ||||
| BC058512.1 | AAH58512.1 |

Genomic Map