Assay Details
Target Gene Details
Entrez Gene ID: | 216846 |
Gene Name: | centrobin, centrosomal BRCA2 interacting protein |
Gene Aliases: |
9830165K03Rik, Lip8 |
Location: |
Chr.11:69299491-69323893 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cntrob | NM_172560.3 | 9 | 2135 | NP_766148.2 |
| AK036710.1 | 8 | 1955 | BAC29545.1 | |
| AK147774.1 | 9 | 2021 | ||
| BC058512.1 | 9 | 968 | AAH58512.1 |

Genomic Map