Assay Details
Target Gene Details
Entrez Gene ID: | 14017 |
Gene Name: | ecotropic viral integration site 2a |
Gene Aliases: |
AW491894, Evi-2, Evi2 |
Location: |
Chr.11:79526560-79530609 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Evi2a | NM_001033711.1 | 2 | 1212 | NP_001028883.1 |
| NM_010161.3 | 2 | 399 | NP_034291.1 | |
| AK005353.1 | 2 | 378 | ||
| AK044098.1 | 2 | 275 | BAC31775.1 | |
| AK046407.1 | 2 | 350 | BAC32711.1 | |
| AK160302.1 | 2 | 316 | BAE35731.1 | |
| AK170527.1 | 2 | 371 | BAE41859.1 | |
| AK171332.1 | 1 | 2747 | BAE42397.1 | |
| AK171594.1 | 2 | 317 | BAE42550.1 | |
| BC038124.1 | 2 | 356 | AAH38124.1 | |
| M34896.1 | 2 | 329 | AAA37583.1 | |
| M34897.1 | 1 | 725 | AAA37584.1 |
Target Gene Details
Entrez Gene ID: | 101488212 |
Gene Name: | predicted gene 21975 |
Gene Aliases: |
Evi2a-evi2b |
Location: |
Chr.11:79513385-79530609 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm21975 | NM_146023.4 | 2 | 399 | NP_666135.1 |
| BB626486.1 | 2 | 350 | ||
| BY214628.1 | 2 | 239 | ||
| BY221230.1 | 2 | 239 |
Target Gene Details
Entrez Gene ID: | 18015 |
Gene Name: | neurofibromatosis 1 |
Gene Aliases: |
AW494271, E030030H24Rik, Nf-1 |
Location: |
Chr.11:79335988-79581609 on Build GRCm38 |
Assay Gene Location: | Within Intron 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nf1 | NM_010897.2 | NP_035027.1 | ||
| L10370.1 | AAA68132.1 |
More Information
Set Membership: |
|

Genomic Map