Assay Details
Target Gene Details
Entrez Gene ID: | 14584 |
Gene Name: | glutamine fructose-6-phosphate transaminase 2 |
Gene Aliases: |
AI480523, GFAT2 |
Location: |
Chr.11:49794155-49838620 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gfpt2 | NM_013529.3 | 12 | 1184 | NP_038557.1 |
| AB016780.1 | 12 | 1150 | BAA74729.1 | |
| AK132821.1 | 11 | 1030 | BAE21379.1 | |
| BC031928.1 | 12 | 1152 | AAH31928.1 |
More Information
Set Membership: |
|

Genomic Map