Assay Details
Target Gene Details
Entrez Gene ID: | 30951 |
Gene Name: | chromobox 8 |
Gene Aliases: |
Pc3 |
Location: |
Chr.11:119038436-119040913 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cbx8 | NM_013926.1 | NP_038954.1 | ||
| AB221673.1 | ||||
| AF180370.1 | AAF25615.1 | |||
| BC014815.1 | AAH14815.1 |
More Information
Set Membership: |
|

Genomic Map