Assay Details
Target Gene Details
Entrez Gene ID: | 20305 |
Gene Name: | chemokine (C-C motif) ligand 6 |
Gene Aliases: |
MRP-1, Scya6, c10 |
Location: |
Chr.11:83587886-83593087 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccl6 | NM_009139.3 | NP_033165.1 | ||
| AF128188.1 | AAF22529.1 | |||
| AF128189.1 | AAF22530.1 | |||
| AF128190.1 | AAF22531.1 | |||
| AF128191.1 | AAF22532.1 | |||
| AF128192.1 | AAF22533.1 | |||
| AK002697.1 | BAB22291.1 | |||
| AK008547.1 | BAB25734.1 | |||
| AK150336.1 | BAE29478.1 | |||
| AK154553.1 | BAE32672.1 | |||
| BC002073.1 | AAH02073.1 | |||
| BY172106.1 | ||||
| M58004.1 | AAA37329.1 |
Target Gene Details
Entrez Gene ID: | 100504464 |
Gene Name: | RIKEN cDNA E230016K23 gene |
Gene Aliases: |
- |
Location: |
Chr.11:83582061-83623693 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E230016K23Rik | NR_036452.1 | |||
| NR_036453.1 | ||||
| AK054074.1 | BAC35646.1 | |||
| AK087588.1 | ||||
| AK154990.1 | BAE32978.1 |
More Information
Set Membership: |
|

Genomic Map