Assay Details
Target Gene Details
Entrez Gene ID: | 13591 |
Gene Name: | early B cell factor 1 |
Gene Aliases: |
Ebf, O/E-1, OE-1, Olf-1, Olf1 |
Location: |
Chr.11:44618100-45008096 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ebf1 | NM_001290709.1 | NP_001277638.1 | ||
| NM_001290710.1 | NP_001277639.1 | |||
| NM_001290711.1 | NP_001277640.1 | |||
| NM_007897.3 | NP_031923.1 | |||
| AK031239.1 | BAC27313.1 | |||
| AK035781.1 | BAC29185.1 | |||
| AK137389.1 | ||||
| AK139021.1 | BAE23864.1 | |||
| AK141175.1 | BAE24573.1 | |||
| BC139362.1 | ||||
| BC139364.1 | ||||
| L12147.1 | AAA37533.1 |
More Information
Set Membership: |
|

Genomic Map