Assay Details
Target Gene Details
Entrez Gene ID: | 208092 |
Gene Name: | charged multivesicular body protein 6 |
Gene Aliases: |
2400004G01Rik |
Location: |
Chr.11:119913446-119919552 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chmp6 | NM_001085498.2 | NP_001078967.1 | ||
| AK162346.1 | BAE36866.1 | |||
| AK172123.1 | ||||
| AK219882.1 | ||||
| BC147754.1 | ||||
| BC147755.1 | ||||
| BG078687.2 |
More Information
Set Membership: |
|

Genomic Map