Assay Details
Target Gene Details
Entrez Gene ID: | 12799 |
Gene Name: | 2',3'-cyclic nucleotide 3' phosphodiesterase |
Gene Aliases: |
CNPase, Cnp-1, Cnp1 |
Location: |
Chr.11:100574909-100591521 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnp |
Target Gene Details
Entrez Gene ID: | 56354 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 7 |
Gene Aliases: |
2010003F24Rik, 2010004G07Rik, CCRP, Ttc2, mDj11, mTpr2 |
Location: |
Chr.11:100582818-100620168 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc7 | NM_019795.4 | NP_062769.2 | ||
| AB028861.1 | BAA88309.1 | |||
| AK008029.1 | ||||
| AK008092.1 | ||||
| AK008268.1 | ||||
| AK011384.1 | BAB27584.1 | |||
| AK011875.1 | BAB27893.1 | |||
| AK076032.1 | BAC36133.1 | |||
| AK145506.1 | BAE26476.1 | |||
| AK145739.1 | BAE26619.1 | |||
| AK166872.1 | BAE39083.1 | |||
| AY323828.1 | AAQ91291.1 | |||
| BC023681.1 | AAH23681.1 | |||
| BC055729.1 | AAH55729.1 |
More Information
Set Membership: |
|

Genomic Map