Assay Details
Target Gene Details
Entrez Gene ID: | 238011 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 7 |
Gene Aliases: |
Alk-SMase, Gm254 |
Location: |
Chr.11:118988108-118993270 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Enpp7 | NM_001030291.1 | NP_001025462.1 | ||
| AK144474.1 | BAE25908.1 | |||
| AK167678.1 | BAE39727.1 | |||
| BC141245.1 | ||||
| BC171967.1 |

Genomic Map