Assay Details
Target Gene Details
Entrez Gene ID: | 14149 |
Gene Name: | ferredoxin reductase |
Gene Aliases: |
- |
Location: |
Chr.11:115268024-115277095 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fdxr | NM_007997.1 | NP_032023.1 | ||
| AK134068.1 | BAE22000.1 | |||
| BC057146.1 | AAH57146.1 | |||
| BC071268.1 | AAH71268.1 | |||
| D49920.1 | BAA08659.1 |
More Information
Set Membership: |
|

Genomic Map