Assay Details
Target Gene Details
Entrez Gene ID: | 52513 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 56 |
Gene Aliases: |
2600001H07Rik, D11Ertd619e, Noh61 |
Location: |
Chr.11:6257545-6267729 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx56 | NM_026538.3 | 3 | 315 | NP_080814.1 |
| AK011136.1 | 3 | 325 | BAB27426.1 | |
| AK076047.1 | 3 | 333 | BAC36145.1 | |
| AK076975.1 | 3 | 315 | BAC36543.1 | |
| AK077514.1 | 3 | 328 | BAC36839.1 | |
| AK145628.1 | 3 | 356 | BAE26549.1 | |
| AK159793.1 | 3 | 319 | BAE35375.1 | |
| AK169654.1 | 3 | 330 | BAE41278.1 | |
| AK188357.1 | 2 | 128 | ||
| AK191115.1 | 2 | 214 | ||
| BC018291.1 | 3 | 309 | AAH18291.1 |

Genomic Map