Assay Details
Target Gene Details
Entrez Gene ID: | 13855 |
Gene Name: | epsin 2 |
Gene Aliases: |
9530051D10Rik, AA536924, Ibp2 |
Location: |
Chr.11:61517249-61579881 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epn2 | NM_001252188.1 | NP_001239117.1 | ||
| NM_001252189.1 | NP_001239118.1 | |||
| NM_010148.3 | NP_034278.1 | |||
| AF057286.1 | AAC97476.1 | |||
| AK036331.1 | BAC29387.1 | |||
| AK139234.1 | ||||
| AK139793.1 | ||||
| AK160782.1 | BAE36006.1 | |||
| AK173093.1 | BAD32371.1 | |||
| BC039138.1 | AAH39138.1 |

Genomic Map