Assay Details
Target Gene Details
Entrez Gene ID: | 67300 |
Gene Name: | clathrin, heavy polypeptide (Hc) |
Gene Aliases: |
3110065L21Rik, CHC, R74732 |
Location: |
Chr.11:86694610-86757742 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cltc | NM_001003908.1 | 25 | 4206 | NP_001003908.1 |
| AK122193.1 | 25 | 4210 | BAC65475.2 | |
| AK143503.1 | 14 | 2097 | BAE25402.1 | |
| BC031408.1 | 4 | 492 | AAH31408.1 | |
| BC079897.1 | 25 | 4206 | AAH79897.1 |
More Information
Set Membership: |
|

Genomic Map