Assay Details
Target Gene Details
Entrez Gene ID: | 67300 |
Gene Name: | clathrin, heavy polypeptide (Hc) |
Gene Aliases: |
3110065L21Rik, CHC, R74732 |
Location: |
Chr.11:86694610-86757742 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cltc | NM_001003908.1 | NP_001003908.1 | ||
| AK122193.1 | BAC65475.2 | |||
| AK143503.1 | BAE25402.1 | |||
| AK167527.1 | BAE39597.1 | |||
| BC031408.1 | AAH31408.1 | |||
| BC079897.1 | AAH79897.1 |
More Information
Set Membership: |
|

Genomic Map