Assay Details
Target Gene Details
Entrez Gene ID: | 104625 |
Gene Name: | CCR4-NOT transcription complex, subunit 6 |
Gene Aliases: |
CCR4 |
Location: |
Chr.11:49671497-49714004 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnot6 | NM_001290741.1 | 9 | 1184 | NP_001277670.1 |
| NM_212484.2 | 10 | 1538 | NP_997649.1 | |
| AK129307.1 | 4 | 2476 | BAC98117.1 | |
| AK212064.1 | ||||
| AK213936.1 | ||||
| AY043269.1 | 9 | 1182 | AAK85707.1 | |
| BC049984.1 | 5 | 708 | AAH49984.1 | |
| BC057190.1 | 4 | 635 | AAH57190.1 | |
| BC062950.1 | 10 | 1526 | AAH62950.1 |
More Information
Set Membership: |
|

Genomic Map