Assay Details
Target Gene Details
Entrez Gene ID: | 57837 |
Gene Name: | Era (G-protein)-like 1 (E. coli) |
Gene Aliases: |
2610524P08Rik, 9130407C09Rik, AU019798, Era, M-ERA, MERA-S, MERA-W |
Location: |
Chr.11:78073376-78080383 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eral1 | NM_022313.2 | 7 | 780 | NP_071708.2 |
| AB049389.1 | 7 | 756 | BAB56113.1 | |
| AF190730.1 | 7 | 748 | AAG17176.1 | |
| AF190731.1 | AAG17177.1 | |||
| AK012155.1 | 7 | 770 | BAB28065.1 | |
| AK153947.1 | 1 | 1039 | ||
| BC019728.1 | AAH19728.1 | |||
| BC068271.1 | 7 | 775 | AAH68271.1 |
More Information
Set Membership: |
|

Genomic Map