Assay Details
Target Gene Details
Entrez Gene ID: | 104625 |
Gene Name: | CCR4-NOT transcription complex, subunit 6 |
Gene Aliases: |
CCR4 |
Location: |
Chr.11:49671497-49714004 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnot6 | NM_001290741.1 | 2 | 178 | NP_001277670.1 |
| NM_212484.2 | 3 | 547 | NP_997649.1 | |
| AK187687.1 | 3 | 194 | ||
| AY043269.1 | 2 | 176 | AAK85707.1 | |
| BC062950.1 | 3 | 535 | AAH62950.1 | |
| BY741392.1 | 3 | 549 | ||
| CF746424.1 |
More Information
Set Membership: |
|

Genomic Map