Assay Details
Target Gene Details
Entrez Gene ID: | 104625 |
Gene Name: | CCR4-NOT transcription complex, subunit 6 |
Gene Aliases: |
CCR4 |
Location: |
Chr.11:49671497-49714004 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnot6 | NM_001290741.1 | NP_001277670.1 | ||
| NM_212484.2 | NP_997649.1 | |||
| AK043052.1 | 1 | 213 | ||
| AK129307.1 | BAC98117.1 | |||
| AY043269.1 | AAK85707.1 | |||
| BC049984.1 | AAH49984.1 | |||
| BC057190.1 | AAH57190.1 | |||
| BC062950.1 | AAH62950.1 | |||
| CF746424.1 |
More Information
Set Membership: |
|

Genomic Map