Assay Details
Target Gene Details
Entrez Gene ID: | 12799 |
Gene Name: | 2',3'-cyclic nucleotide 3' phosphodiesterase |
Gene Aliases: |
CNPase, Cnp-1, Cnp1 |
Location: |
Chr.11:100574909-100591521 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnp | NM_009923.2 | NP_034053.2 | ||
| AK050628.1 | BAC34351.1 | |||
| AK087415.1 | ||||
| AK154654.1 | 1 | 89 | BAE32745.1 | |
| AK158318.1 | BAE34457.1 | |||
| AK158520.1 | BAE34543.1 | |||
| AK172318.1 | ||||
| AK172514.1 | ||||
| BC005544.1 | AAH05544.1 | |||
| BY333311.1 | ||||
| M58045.1 | AAA37430.1 |
More Information
Set Membership: |
|

Genomic Map