Assay Details
Target Gene Details
Entrez Gene ID: | 68460 |
Gene Name: | dehydrogenase/reductase (SDR family) member 7C |
Gene Aliases: |
1110001P11Rik, AI120487, SRP-35 |
Location: |
Chr.11:67798271-67816002 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhrs7c | NM_001013013.2 | NP_001013031.2 | ||
| AI391290.1 |

Genomic Map