Assay Details
Target Gene Details
Entrez Gene ID: | 66569 |
Gene Name: | glycerophosphodiester phosphodiesterase domain containing 1 |
Gene Aliases: |
2610020H15Rik |
Location: |
Chr.11:87033794-87074953 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gdpd1 | NM_025638.2 | NP_079914.1 | ||
| AK011487.1 | BAB27650.1 | |||
| AK013864.1 | BAB29022.1 | |||
| AK016023.1 | BAB30083.1 | |||
| AK167812.1 | BAE39839.1 | |||
| BC016541.1 | AAH16541.1 |
More Information
Set Membership: |
|

Genomic Map