Assay Details
Target Gene Details
Entrez Gene ID: | 14257 |
Gene Name: | FMS-like tyrosine kinase 4 |
Gene Aliases: |
AI323512, Chy, Flt-4, VEGFR-3, VEGFR3 |
Location: |
Chr.11:49609679-49652739 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Flt4 | NM_008029.3 | 10 | 1395 | NP_032055.1 |
| BC138346.1 | 10 | 1367 | ||
| BC138348.1 | 10 | 1367 | ||
| L07296.1 | 10 | 1348 | AAA40077.1 |
More Information
Set Membership: |
|

Genomic Map