Assay Details
Target Gene Details
Entrez Gene ID: | 16897 |
Gene Name: | lethal giant larvae homolog 1 (Drosophila) |
Gene Aliases: |
AI325176, Lgl1, Llglh, Mgl1 |
Location: |
Chr.11:60699670-60714188 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Llgl1 | NM_001159404.1 | 21 | 3218 | NP_001152876.1 |
| NM_001159405.1 | 21 | 3218 | NP_001152877.1 | |
| NM_008502.2 | 21 | 3212 | NP_032528.1 | |
| AK083672.1 | 5 | 798 | BAC38989.1 | |
| AK084386.1 | 13 | 2088 | BAC39171.1 | |
| AK159412.1 | 21 | 3084 | BAE35062.1 | |
| AK184080.1 | 1 | 39 | ||
| BC043458.1 | 7 | 1123 | AAH43458.1 | |
| BC050913.1 | 21 | 3063 | AAH50913.1 | |
| BC055399.1 | 21 | 3185 | AAH55399.1 | |
| D16141.1 | 21 | 3062 | BAA03712.1 |
More Information
Set Membership: |
|

Genomic Map