Assay Details
Target Gene Details
Entrez Gene ID: | 17885 |
Gene Name: | myosin, heavy polypeptide 8, skeletal muscle, perinatal |
Gene Aliases: |
4832426G23Rik, AI327267, MHCp, MyHC-pn, Myhs-p, Myhsp |
Location: |
Chr.11:67277124-67308634 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myh8 | NM_177369.3 | NP_796343.2 | ||
| AK029328.1 | ||||
| AK220548.1 | BAD90320.1 | |||
| BC150737.1 |

Genomic Map