Assay Details
Target Gene Details
Entrez Gene ID: | 217127 |
Gene Name: | K(lysine) acetyltransferase 7 |
Gene Aliases: |
Hbo1, Hboa, Myst2 |
Location: |
Chr.11:95271853-95310246 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kat7 | NM_001195003.1 | NP_001181932.1 | ||
| NM_001195004.1 | NP_001181933.1 | |||
| NM_177619.3 | NP_808287.1 | |||
| AK078483.1 | ||||
| AK147968.1 | BAE28257.1 | |||
| AK164336.1 | 1 | 4118 | ||
| AK169458.1 | ||||
| AK170389.1 | BAE41763.1 | |||
| AK180889.1 | ||||
| AK181682.1 | ||||
| AK216566.1 | ||||
| BC048904.1 | AAH48904.1 | |||
| BC057102.1 | AAH57102.1 | |||
| CD555481.1 | ||||
| DQ076247.1 |
More Information
Set Membership: |
|

Genomic Map