Assay Details
Target Gene Details
Entrez Gene ID: | 56494 |
Gene Name: | golgi SNAP receptor complex member 2 |
Gene Aliases: |
2310032N09Rik, C76855, Gs27, SNARE, membrin |
Location: |
Chr.11:103676849-103697738 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gosr2 | NM_019650.3 | NP_062624.2 | ||
| AF007550.1 | AAB82653.1 | |||
| AK002556.1 | ||||
| AK005697.1 | BAB24194.1 | |||
| AK013052.1 | BAB28622.1 | |||
| AK150004.1 | BAE29231.1 | |||
| BC008525.1 | AAH08525.1 | |||
| BC051253.1 | AAH51253.1 |
More Information
Set Membership: |
|

Genomic Map