Assay Details
Target Gene Details
Entrez Gene ID: | 170835 |
Gene Name: | inositol polyphosphate 5-phosphatase J |
Gene Aliases: |
Pib5pa, Pipp |
Location: |
Chr.11:3494272-3505567 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inpp5j | NM_172439.3 | NP_766027.2 | ||
| AK034272.1 | BAC28654.1 | |||
| AK136908.1 | ||||
| AK165718.1 | 8 | 2775 | ||
| AK170114.1 | BAE41572.1 | |||
| AK171029.1 | BAE42196.1 | |||
| BC022915.1 | ||||
| BC131634.1 | ||||
| BC131635.1 |
More Information
Set Membership: |
|

Genomic Map