Assay Details
Target Gene Details
Entrez Gene ID: | 60441 |
Gene Name: | mitochondrial ribosomal protein L38 |
Gene Aliases: |
1110036N21Rik, 4733401F03Rik, MRP-L3, R75240, Rpml3 |
Location: |
Chr.11:116131817-116138868 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl38 | NM_024177.3 | 2 | 249 | NP_077139.2 |
| AK010798.1 | 2 | 193 | ||
| AK013126.1 | 2 | 216 | ||
| AK014632.1 | 2 | 245 | ||
| AK088343.1 | 2 | 188 | BAC40295.1 | |
| AK151598.1 | 2 | 188 | BAE30538.1 | |
| AK159033.1 | 2 | 250 | BAE34778.1 | |
| BC002319.1 | 1 | 225 | AAH02319.1 | |
| BC030739.1 | 2 | 176 | AAH30739.1 |
More Information
Set Membership: |
|

Genomic Map