Assay Details
Target Gene Details
Entrez Gene ID: | 76113 |
Gene Name: | lactoperoxidase |
Gene Aliases: |
5830499B15Rik |
Location: |
Chr.11:87806428-87826135 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lpo | NM_080420.2 | NP_536345.2 | ||
| AK018070.1 | ||||
| AK162992.1 | BAE37148.1 | |||
| BC016212.1 | AAH16212.1 |
More Information
Set Membership: |
|

Genomic Map