Assay Details
Target Gene Details
Entrez Gene ID: | 20185 |
Gene Name: | nuclear receptor co-repressor 1 |
Gene Aliases: |
5730405M06Rik, A230020K14Rik, N-CoR, RIP13, Rxrip13, mKIAA1047 |
Location: |
Chr.11:62315643-62457332 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncor1 | NM_001252313.1 | 26 | 3693 | NP_001239242.1 |
| NM_011308.3 | 26 | 3531 | NP_035438.3 | |
| AB093281.1 | 9 | 1571 | BAC41465.1 | |
| AK137695.1 | 17 | 2569 | BAE23465.1 | |
| BC086657.1 | 26 | 3497 | AAH86657.1 | |
| L78294.1 | 6 | 638 | AAL40135.1 | |
| U35312.1 | 25 | 3736 | AAB17125.1 |
More Information
Set Membership: |
|

Genomic Map