Assay Details
Target Gene Details
Entrez Gene ID: | 14778 |
Gene Name: | glutathione peroxidase 3 |
Gene Aliases: |
AA960521, EGPx, GPx, GSHPx-3, GSHPx-P |
Location: |
Chr.11:54902854-54910382 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpx3 | NM_008161.3 | NP_032187.2 | ||
| AK002219.1 | BAC55243.1 | |||
| AK002262.1 | ||||
| AK004942.1 | BAC55250.1 | |||
| AK146760.1 | BAE27413.1 | |||
| BC003339.1 | AAH03339.1 | |||
| BC037027.1 | AAH37027.1 | |||
| BC049235.1 | AAH49235.1 | |||
| BC061950.1 | AAH61950.1 | |||
| BY061328.1 | ||||
| CB601893.1 | ||||
| U13705.1 | AAA62283.2 |
More Information
Set Membership: |
|

Genomic Map