Assay Details
Target Gene Details
Entrez Gene ID: | 16399 |
Gene Name: | integrin alpha 2b |
Gene Aliases: |
AI172977, CD41, CD41B, GpIIb, alphaIIb |
Location: |
Chr.11:102453297-102469883 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itga2b | NM_010575.2 | NP_034705.2 | ||
| AF170316.1 | AAD56216.1 | |||
| AK048700.1 | BAC33425.1 | |||
| AK140749.1 | ||||
| AK142289.1 | BAE25013.1 | |||
| AK154619.1 | BAE32718.1 | |||
| AK156872.1 | ||||
| AK157012.1 | ||||
| AK158443.1 | ||||
| BC120493.1 | ||||
| BC137570.1 |
More Information
Set Membership: |
|

Genomic Map