Assay Details
Target Gene Details
Entrez Gene ID: | 16428 |
Gene Name: | IL2 inducible T cell kinase |
Gene Aliases: |
Emt, Tcsk, Tsk |
Location: |
Chr.11:46325148-46389515 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itk | NM_001281965.1 | NP_001268894.1 | ||
| NM_001281966.1 | NP_001268895.1 | |||
| NM_001281967.1 | NP_001268896.1 | |||
| NM_001281968.1 | NP_001268897.1 | |||
| NM_010583.3 | NP_034713.2 | |||
| AK079693.1 | ||||
| AK083380.1 | ||||
| AK153613.1 | BAE32118.1 | |||
| BC128374.1 | ||||
| BC128375.1 | ||||
| D14042.1 | BAA03129.1 | |||
| L00619.1 | AAA39337.1 | |||
| L05631.1 | AAA40518.1 | |||
| L10628.1 |
More Information
Set Membership: |
|

Genomic Map