Assay Details
Target Gene Details
Entrez Gene ID: | 16480 |
Gene Name: | junction plakoglobin |
Gene Aliases: |
Ctnng, D930025P04Rik, PG |
Location: |
Chr.11:100368856-100397790 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jup | NM_010593.2 | NP_034723.1 | ||
| AK086401.1 | 1 | 52 | ||
| AK145895.1 | ||||
| AK170934.1 | BAE42126.1 | |||
| BC040757.1 | AAH40757.1 | |||
| BC094461.1 | AAH94461.1 | |||
| M90365.1 | AAB02885.1 |
More Information
Set Membership: |
|

Genomic Map