Assay Details
Target Gene Details
Entrez Gene ID: | 76784 |
Gene Name: | mitochondrial translational initiation factor 2 |
Gene Aliases: |
2310038D14Rik, 2410112O06Rik, IF-2(Mt), IF-2mt, IF2(mt) |
Location: |
Chr.11:29526379-29545452 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtif2 | NM_001282118.1 | 3 | 458 | NP_001269047.1 |
| NM_001282119.1 | 3 | 380 | NP_001269048.1 | |
| NM_001282120.1 | 3 | 736 | NP_001269049.1 | |
| NM_133767.3 | 3 | 736 | NP_598528.2 | |
| AK010766.1 | 3 | 681 | ||
| AK136344.1 | 3 | 420 | BAE22940.1 | |
| AK146961.1 | 3 | 676 | BAE27568.1 | |
| AK160724.1 | ||||
| BC016590.1 | 3 | 312 | AAH16590.1 | |
| CK620846.1 | 3 | 369 |
More Information
Set Membership: |
|

Genomic Map