Assay Details
Target Gene Details
Entrez Gene ID: | 14784 |
Gene Name: | growth factor receptor bound protein 2 |
Gene Aliases: |
AA408164, Ash |
Location: |
Chr.11:115644130-115708597 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grb2 | NM_008163.3 | NP_032189.1 | ||
| AK077750.1 | ||||
| AK079087.1 | ||||
| AK148148.1 | BAE28376.1 | |||
| AK153557.1 | BAE32093.1 | |||
| AK155800.1 | BAE33439.1 | |||
| AK166142.1 | BAE38595.1 | |||
| AK170845.1 | BAE42068.1 | |||
| AK172002.1 | ||||
| AK196759.1 | ||||
| BC052377.1 | AAH52377.1 | |||
| BC085254.1 | AAH85254.1 | |||
| D85748.1 | BAA12862.1 | |||
| U07617.1 | AAB40022.1 |

Genomic Map