Assay Details
Target Gene Details
Entrez Gene ID: | 380684 |
Gene Name: | neurofilament, heavy polypeptide |
Gene Aliases: |
NF-H, NF200, Nfh, mKIAA0845 |
Location: |
Chr.11:4938754-4948064 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nefh | NM_010904.3 | 2 | 1150 | NP_035034.2 |
| AK122388.1 | 2 | 1052 | BAC65670.1 | |
| M35131.1 | 3 | 1173 | AAA39809.1 |

Genomic Map