Assay Details
Target Gene Details
Entrez Gene ID: | 14534 |
Gene Name: | K(lysine) acetyltransferase 2A |
Gene Aliases: |
1110051E14Rik, AW212720, Gcn5, Gcn5l2, mmGCN5 |
Location: |
Chr.11:100704746-100712467 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kat2a | NM_001038010.2 | 8 | 1278 | NP_001033099.1 |
| NM_020004.5 | 8 | 1278 | NP_064388.2 | |
| AF254441.1 | 8 | 1215 | AAF70497.1 | |
| AK137358.1 | 8 | 1255 | BAE23321.1 | |
| AK145205.1 | 7 | 1176 | BAE26296.1 | |
| AK147042.1 | 8 | 1226 | ||
| AK150225.1 | 8 | 1256 | BAE29392.1 | |
| AK154866.1 | 8 | 1266 | ||
| AK155347.1 | 8 | 1455 | BAE33207.1 | |
| AK155791.1 | 8 | 1280 | ||
| AK155842.1 | 8 | 1279 | BAE33458.1 | |
| AK158079.1 | 8 | 1266 | BAE34351.1 | |
| BC063752.1 | 8 | 1263 | AAH63752.1 |

Genomic Map